Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl Cholecystokinin, CCK (26-31), sulfated

Chemical Identifiers

Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31)
Name Note sulfated
Formula Weight 923.04
MDL Number MFCD00133564
Synonym N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-Met-NH2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66989EXD

 
alfa aesar™
J66989EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66989MCR

 
alfa aesar™
J66989MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 923.04
Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31)
MDL Number MFCD00133564
Name Note sulfated
Physical Form Solid
Synonym N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-Met-NH2
Color White to off-white
Storage Note 1 -30° to -10°C
Quantity 0.5mg
Safety and Handling

-30° to -10°C