Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl Cholecystokinin, CCK (26-31), sulfated


Quantity 1mg

Manufacturer: alfa aesar™  J66989MCR

 View more versions of this product

Catalog No. AAJ66989MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 923.04
Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31)
MDL Number MFCD00133564
Synonym N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-Met-NH2
Name Note sulfated
Physical Form Solid
Color White to off-white
Quantity 1mg
Storage Note 1 -30° to -10°C

Specifications

Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31)
Name Note sulfated
Formula Weight 923.04
MDL Number MFCD00133564
Synonym N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-Met-NH2
Accessories

-30° to -10°C