Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl Cholecystokinin, CCK (26-31), non-sulfated

Chemical Identifiers

Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31)
Name Note non-sulfated
Formula Weight 842.98
MDL Number MFCD00133562
Synonym N-Acetyl-Asp-Tyr-Met-Gly-Trp-Met-NH2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66976EXD

 
alfa aesar™
J66976EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66976MCR

 
alfa aesar™
J66976MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 842.98
Chemical Name or Material N-Acetyl Cholecystokinin, CCK (26-31)
Physical Form Solid
MDL Number MFCD00133562
Synonym N-Acetyl-Asp-Tyr-Met-Gly-Trp-Met-NH2
Storage Note 1 -30° to -10°C
Color White to off-white
Quantity 0.5mg
Name Note non-sulfated
Safety and Handling

-30° to -10°C