missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl Cholecystokinin, CCK (26-30), sulfated

Chemical Identifiers

MDL Number MFCD00133563
Synonym N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-NH2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66056EXD
View Documents
Alfa Aesar™
J66056EXD
0.5mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66056MCR
View Documents
Alfa Aesar™
J66056MCR
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

MFCD00133563
N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-NH2

Specifications

White
MFCD00133563
791.85
N-Acetyl Cholecystokinin, CCK (26-30), sulfated
0.5mg
N-Acetyl-Asp-Tyr(SO{3}H)-Met-Gly-Trp-NH2
Solid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS