Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Asp-Tyr(PO_3H_2)-Val-Pro-Met-Leu-NH_2


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66744MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66744MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1mg
Format Solid
Sequence Ac-Asp-Tyr(PO3H2)-Val-Pro-Met-Leu-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type N-Acetyl-Asp-Tyr(PO_3H_2)-Val-Pro-Met-Leu-NH_2
Molecular Weight 857.91
Purification Method HPLC