Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH_2

For cellular and molecular biology applications

$120.54 - $183.82

Specifications

Product Type N-Acetyl-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH_2
Molecular Weight 879.98
Storage Requirements -30° to -10°C
Sequence Ac-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66171EXD

 
alfa aesar™
J66171EXD
0.5mg Each for $120.54

AAJ66171MCR

 
alfa aesar™
J66171MCR
1mg Each for $183.82
Description & Specifications

Specifications

Product Type N-Acetyl-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH_2
Molecular Weight 879.98
Storage Requirements -30° to -10°C
Sequence Ac-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC