Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl Angiotensinogen (1-14), Human

For cellular and molecular biology applications

Specifications

Product Type N-Acetyl Angiotensinogen (1-14), human
Cross Reactivity Human
Molecular Weight 1802.07
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Asn
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66284LB0

 
alfa aesar™
J66284LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66284MCR

 
alfa aesar™
J66284MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type N-Acetyl Angiotensinogen (1-14), human
Cross Reactivity Human
Molecular Weight 1802.07
Storage Requirements -30° to -10°C
Sequence N-Acetyl-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Asn
Format Solid
Regulatory Status RUO
Purification Method HPLC