Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Adhesin (1025-1044) amide

Chemical Identifiers

Chemical Name or Material N-Acetyl-Adhesin (1025-1044) amide
Formula Weight 2202.46
MDL Number MFCD02262418
Synonym N-Acetyl-Gln-Leu-Lys-Thr-Ala-Asp-Leu-Pro-Ala-Gly-Arg-Asp-Glu-Thr-Thr-Ser-Phe-Val-Leu-Val-NH2; p1025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66492EXD

 
alfa aesar™
J66492EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66492MCR

 
alfa aesar™
J66492MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 2202.46
MDL Number MFCD02262418
Synonym N-Acetyl-Gln-Leu-Lys-Thr-Ala-Asp-Leu-Pro-Ala-Gly-Arg-Asp-Glu-Thr-Thr-Ser-Phe-Val-Leu-Val-NH2; p1025
Physical Form Lyophilized Powder
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Chemical Name or Material N-Acetyl-Adhesin (1025-1044) amide
Safety and Handling

-30° to -10°C