Please login to your online account to display your discounted pricing

N-Acetyl-5-hydroxytryptamine, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  226932500

 View more versions of this product

Catalog No. AC226932500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Appearance of Solution (5% soln. in EtOH) clear, colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Water 1% max. (K.F.)
Color White to off-white or slightly pink
Quantity 250mg
Chemical Name or Material N-Acetyl-5-hydroxytryptamine
Name Note 99%
CAS 1210-83-9
CAS Min % 98.5
CAS Max % 100.0
Physical Form White to off-white or slightly pink powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C12H14N2O2
Formula Weight 218.25g/mol
MDL Number MFCD00005656
Beilstein 22, V,12, 41
Synonym N-Acetylserotonin

Specifications

Chemical Name or Material N-Acetyl-5-hydroxytryptamine
Name Note 99%
CAS 1210-83-9
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C12H14N2O2
Formula Weight 218.25g/mol
MDL Number MFCD00005656
Synonym N-Acetylserotonin