Please login to your online account to display your discounted pricing

N-Acetyl-5-hydroxytryptamine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Acetyl-5-hydroxytryptamine
Name Note 99%
CAS 1210-83-9
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C12H14N2O2
Formula Weight 218.25g/mol
MDL Number MFCD00005656
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC226932500

  SDS
thermo fisher scientific
226932500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Appearance of Solution (5% soln. in EtOH) clear, colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Water 1% max. (K.F.)
Color White to off-white or slightly pink
Quantity 250mg
Chemical Name or Material N-Acetyl-5-hydroxytryptamine
Name Note 99%
CAS 1210-83-9
CAS Min % 98.5
CAS Max % 100.0
Physical Form White to off-white or slightly pink powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C12H14N2O2
Formula Weight 218.25g/mol
MDL Number MFCD00005656
Beilstein 22, V,12, 41
Synonym N-Acetylserotonin