missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-4-piperidone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166820250

 View more versions of this product

Catalog No. AC166820250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Acetyl-4-piperidone
>112°C
Glass bottle
1.14
Orange
97%
96.0
96% min. (GC)
MFCD00006190
NNFOVLFUGLWWCL-UHFFFAOYSA-N
1-acetylpiperidin-4-one
122563
Oily Liquid
1.1400g/mL
Authentic
1.5015 to 1.5035
218°C
25g
32161-06-1
100.0
C7H11NO2
1-acetyl-4-oxopiperidine
CC(=O)N1CCC(=O)CC1
141.17
141.17
97%

Chemical Identifiers

32161-06-1
141.17
NNFOVLFUGLWWCL-UHFFFAOYSA-N
122563
CC(=O)N1CCC(=O)CC1
C7H11NO2
MFCD00006190
1-acetyl-4-oxopiperidine
1-acetylpiperidin-4-one
Safety and Handling

Safety and Handling

EINECSNumber : 250-937-6

SDS Documents