missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-4-piperidone, 97%, ACROS Organics™

Chemical Identifiers

CAS 32161-06-1
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H11NO2
Formula Weight 141.17
MDL Number MFCD00006190
Chemical Name or Material N-Acetyl-4-piperidone, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC166820250 SDS Fisher Scientific Acros
166820250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC166821000 SDS Fisher Scientific Acros
166821000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

32161-06-1
100.0
C7H11NO2
MFCD00006190
≥96.0%
96.0
97%
141.17
N-Acetyl-4-piperidone, 97%

Specifications

141.17
25g
96.0
1.5015 to 1.5035 (20°C, 589nm)
C7H11NO2
97%
Authentic
≥96.0%
Oily Liquid
Clear orange
100.0
32161-06-1
MFCD00006190
Glass bottle
N-Acetyl-4-piperidone, 97%

Safety and Handling

EINECSNumber : 250-937-6