Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-3-(2-naphthyl)-D-alanine, 95%

Chemical Identifiers

Chemical Name or Material N-Acetyl-3-(2-naphthyl)-D-alanine
CAS 37440-01-0
Formula Weight 257.29
MDL Number MFCD00079678
Beilstein 7193723
Synonym Ac-D-Ala(2-naphthyl)-OH; (R)-2-Acetyl-3-(2-naphthyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6619903

 
alfa aesar™
H6619903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66199MD

 
alfa aesar™
H66199MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6619906

 
alfa aesar™
H6619906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 257.29
Chemical Name or Material N-Acetyl-3-(2-naphthyl)-D-alanine
MDL Number MFCD00079678
Physical Form Powder
Assay 95%
CAS 37440-01-0
Beilstein 7193723
Synonym Ac-D-Ala(2-naphthyl)-OH; (R)-2-Acetyl-3-(2-naphthyl)propionic acid
Color Off-white to white
Storage Note 1 -30° to -10°C
Quantity 1g
Safety and Handling

-30° to -10°C