missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-2-phenylethylamine, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 243021000

 View more versions of this product

Catalog No. AC243021000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
163.22
White to off-white
100.0
MFCD00026177
98%
N-Acetyl-2-phenylethylamine
877-95-2
Low Melting Mass
100g
49° to 55°C
Authentic
97.5
98%
C10H13NO
C6H5CH2CH2NHCOCH3

Chemical Identifiers

877-95-2
100.0
C10H13NO
163.22
N-Acetyl-2-phenylethylamine, 98%
97.5
98%
C6H5CH2CH2NHCOCH3
MFCD00026177
≥97.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 212-897-8

RTECSNumber : AC7740000