Please login to your online account to display your discounted pricing

N-Acetyl-2-phenylethylamine, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material N-Acetyl-2-phenylethylamine
Name Note 98%
CAS 877-95-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H13NO
Linear Formula C6H5CH2CH2NHCOCH3
Formula Weight 163.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC243021000

  SDS
thermo fisher scientific
243021000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Low Melting Mass
Formula Weight 163.22
Quantity 100g
Color White to off-white
Melting Point 49° to 55°C
EINECS Number 212-897-8
RTECS Number AC7740000
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00026177
CAS Min % 97.5
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material N-Acetyl-2-phenylethylamine
Molecular Formula C10H13NO
CAS 877-95-2
Linear Formula C6H5CH2CH2NHCOCH3