Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetonylpyridinium bromide, 90+%

$34.49 - $423.30

Chemical Identifiers

Chemical Name or Material N-Acetonylpyridinium bromide
CAS 17282-41-6
Molecular Formula C8H10BrNO
Formula Weight 216.08
MDL Number MFCD00031948
Beilstein 3574011
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2449004

 
alfa aesar™
B2449004
2g Each for $34.49

AAB2449018

 
alfa aesar™
B2449018
50g Each for $423.30
Description & Specifications

Specifications

Formula Weight 216.08
Chemical Name or Material N-Acetonylpyridinium bromide
Assay ≥90%
Molecular Formula C8H10BrNO
MDL Number MFCD00031948
CAS 17282-41-6
Beilstein 3574011
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈185°C
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.