missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]succinimide 98.0+%, TCI America™
Supplier: TCI America F023925G
Specifications
N-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]succinimide | |
153°C | |
C19H15NO5 | |
25 g | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate | |
134122 | |
≥98.0% (HPLC,T) |
82911-69-1 | |
White | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
337.331 | |
337.33 | |
Crystalline Powder |
Chemical Identifiers
82911-69-1 | |
337.331 | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
134122 | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C19H15NO5 | |
MFCD00010733 | |
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator