missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine 98.0+%, TCI America™
Supplier: TCI America F029725G
Specifications
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine | |
183°C | |
C24H21NO4 | |
25 g | |
SJVFAHZPLIXNDH-QFIPXVFZSA-N | |
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid | |
978331 | |
≥98.0% (HPLC,T) |
35661-40-6 | |
White | |
MFCD00037128 | |
fmoc-l-phenylalanine, fmoc-phe-oh, n-fmoc-l-phenylalanine, n-9-fmoc-l-phenylglycine, n-9-fluorenylmethoxycarbonyl-l-phenylalanine, fmoc-l-phe-oh, fmoc-phe, fmoc-l-phe, fmoc-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalanine | |
OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
387.44 | |
387.44 | |
Crystalline Powder |
Chemical Identifiers
35661-40-6 | |
387.44 | |
SJVFAHZPLIXNDH-QFIPXVFZSA-N | |
978331 | |
OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C24H21NO4 | |
MFCD00037128 | |
fmoc-l-phenylalanine, fmoc-phe-oh, n-fmoc-l-phenylalanine, n-9-fmoc-l-phenylglycine, n-9-fluorenylmethoxycarbonyl-l-phenylalanine, fmoc-l-phe-oh, fmoc-phe, fmoc-l-phe, fmoc-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalanine | |
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid |
Safety and Handling
TSCA : No