Learn More
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC243100010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide | |
97.5 | |
(5% soln.in dichloroethane) clear and colorless | |
145.0°C to 153.0°C | |
97.5% min. (HPLC) | |
C19H15NO5 | |
Glass bottle | |
1g | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate | |
134122 | |
Microfine Powder |
82911-69-1 | |
100.0 | |
White | |
0.25% max. (60°C, 2 hrs, 1mm Hg) | |
Authentic | |
MFCD00010733 | |
9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethyl succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, fmoc n-hydroxysuccinimide ester, fmoc-onsu, fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide | |
Solubility in water: insoluble | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
337.29 | |
337.29 | |
98% |
Chemical Identifiers
82911-69-1 | |
337.29 | |
WMSUFWLPZLCIHP-UHFFFAOYSA-N | |
134122 | |
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C19H15NO5 | |
MFCD00010733 | |
9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethyl succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, fmoc n-hydroxysuccinimide ester, fmoc-onsu, fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide | |
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only