Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(5-Nitro-2-pyridyl)-1,2-ethanediamine, 99%

Chemical Identifiers

Chemical Name or Material N-(5-Nitro-2-pyridyl)-1,2-ethanediamine
CAS 29602-39-9
Molecular Formula C7H10N4O2
Formula Weight 182.18
MDL Number MFCD00006254
UN Number UN3259
Synonym 2-(2-Aminoethylamino)-5-nitropyridine; 5-Nitro-2-(2-pyridylaminoethylamine)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2854109

  SDS
alfa aesar™
H2854109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.18
Melting Point 126° to 128°C
Molecular Formula C7H10N4O2
Chemical Name or Material N-(5-Nitro-2-pyridyl)-1,2-ethanediamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN3259
Assay 99%
CAS 29602-39-9
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Quantity 10g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
MDL Number MFCD00006254
Synonym 2-(2-Aminoethylamino)-5-nitropyridine; 5-Nitro-2-(2-pyridylaminoethylamine)
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.