Please login to your online account to display your discounted pricing

N-[(5-Methoxy-1H-indol-3-yl)methyl]-N,N-dimethylamine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Methoxygramine
CAS 16620-52-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12 H16 N2 O
Formula Weight 204.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC125860010

  SDS
fisher scientific acros
125860010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 204.27
Quantity 1g
Color White to almost white
Melting Point 125° to 127°C
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Chemical Name or Material 5-Methoxygramine
Molecular Formula C12 H16 N2 O
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 16620-52-3
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.