missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(5-Hexynyl)phthalimide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5348914

 View more versions of this product

Catalog No. AAH5348914



Description

Description

6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

N-(5-Hexynyl)phthalimide
68°C to 72°C
MFCD00671372
n-5-hexynyl phthalimide, 2-hex-5-yn-1-yl isoindoline-1,3-dione, 6-phthalimido-1-hexyne, 2-hex-5-yn-1-yl isoindole-1,3-dione, acmc-20akav, n-5-hexynylphthalimide, n-hex-5-ynylphthalimide, 6-phthalimidyl-1-hexyne
C#CCCCCN1C(=O)C2=CC=CC=C2C1=O
227.26
227.26
6097-08-1
C14H13NO2
25 g
WALVSJIWVWDFCU-UHFFFAOYSA-N
2-hex-5-ynylisoindole-1,3-dione
3346053
97%

Chemical Identifiers

6097-08-1
MFCD00671372
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only