missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(5-Fluoro-2-methylphenyl)thiourea, 97%, Thermo Scientific™

$39.00 - $154.53

Chemical Identifiers

CAS 16822-86-9
Molecular Formula C8H9FN2S
Molecular Weight (g/mol) 184.23
MDL Number MFCD00052380
InChI Key NZTMKTPFNGHTJG-UHFFFAOYSA-N
Synonym 1-5-fluoranyl-2-methyl-phenyl thiourea, 1-5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl-2-thiourea, 5-fluoro-2-methyl-phenyl thiourea, 5-fluoro-2-methyl-phenyl-thiourea, 5-fluoro-2-methylphenyl thiourea, acmc-1c5tk, n-5-fluoro-2-methylphenyl thiourea
PubChem CID 2732698
IUPAC Name (5-fluoro-2-methylphenyl)thiourea
SMILES CC1=CC=C(F)C=C1NC(N)=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2042303
View Documents Promotion Details
Thermo Fisher Scientific
B2042303
1g
Each for $39.00
 
AAB2042306
View Documents Promotion Details
Thermo Fisher Scientific
B2042306
5g
Each for $154.53
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16822-86-9
184.23
NZTMKTPFNGHTJG-UHFFFAOYSA-N
2732698
CC1=CC=C(F)C=C1NC(N)=S
C8H9FN2S
MFCD00052380
1-5-fluoranyl-2-methyl-phenyl thiourea, 1-5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl-2-thiourea, 5-fluoro-2-methyl-phenyl thiourea, 5-fluoro-2-methyl-phenyl-thiourea, 5-fluoro-2-methylphenyl thiourea, acmc-1c5tk, n-5-fluoro-2-methylphenyl thiourea
(5-fluoro-2-methylphenyl)thiourea

Specifications

16822-86-9
C8H9FN2S
UN2811
1g
NZTMKTPFNGHTJG-UHFFFAOYSA-N
(5-fluoro-2-methylphenyl)thiourea
2732698
97%
117°C to 120°C
MFCD00052380
Odorless
1-5-fluoranyl-2-methyl-phenyl thiourea, 1-5-fluoro-2-methylphenyl thiourea, 1-5-fluoro-2-methylphenyl-2-thiourea, 5-fluoro-2-methyl-phenyl thiourea, 5-fluoro-2-methyl-phenyl-thiourea, 5-fluoro-2-methylphenyl thiourea, acmc-1c5tk, n-5-fluoro-2-methylphenyl thiourea
CC1=CC=C(F)C=C1NC(N)=S
184.23
184.23
N-(5-Fluoro-2-methylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications