Please login to your online account to display your discounted pricing

N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  ACAC378800010

 View more versions of this product

Catalog No. AC378800010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 392.69
Quantity 1g
Chemical Name or Material N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
Packaging Glass bottle
Molecular Formula C7H3ClF6N2O4S2
MDL Number MFCD00191833
CAS 145100-51-2
Synonym 2-[N, N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)
CAS 145100-51-2
Molecular Formula C7H3ClF6N2O4S2
Formula Weight 392.69
MDL Number MFCD00191833
Synonym 2-[N, N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents