Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(5-Chloro-2-methylphenyl)maleamic acid, 97%

$31.95 - $286.28

Chemical Identifiers

Chemical Name or Material N-(5-Chloro-2-methylphenyl)maleamic acid
CAS 208663-08-5
Molecular Formula C11H10ClNO3
Formula Weight 239.64
MDL Number MFCD00135077
Synonym 4-(5-Chloro-2-methylanilino)-4-oxobut-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2489703

  SDS
alfa aesar™
B2489703
1g Each for $31.95

AAB2489706

  SDS
alfa aesar™
B2489706
5g Each for $85.58

AAB2489714

  SDS
alfa aesar™
B2489714
25g Each for $286.28
Description & Specifications

Specifications

Formula Weight 239.64
Chemical Name or Material N-(5-Chloro-2-methylphenyl)maleamic acid
Assay 97%
Molecular Formula C11H10ClNO3
Melting Point 118° to 121°C
MDL Number MFCD00135077
CAS 208663-08-5
Quantity 1g
Synonym 4-(5-Chloro-2-methylanilino)-4-oxobut-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.