Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(5-Chloro-2-methoxyphenyl)thiourea, 98%

$73.08 - $304.36

Chemical Identifiers

Chemical Name or Material N-(5-Chloro-2-methoxyphenyl)thiourea
CAS 63980-69-8
Molecular Formula C8H9ClN2OS
Formula Weight 216.69
MDL Number MFCD00041169
Beilstein 2840488
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1355706

  SDS
alfa aesar™
A1355706
5g Each for $73.08

AAA1355714

  SDS
alfa aesar™
A1355714
25g Each for $304.36
Description & Specifications

Specifications

Formula Weight 216.69
Quantity 5g
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Melting Point 144° to 148°C
Chemical Name or Material N-(5-Chloro-2-methoxyphenyl)thiourea
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Beilstein 2840488
MDL Number MFCD00041169
CAS 63980-69-8
Molecular Formula C8H9ClN2OS
Health Hazard 1 Warning
Assay 98%
RTECS Number YS5950000
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.