missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(5-Anilino-2,4-pentadienylidene)anilinehydrochloride 98%, ACROS Organics™

Chemical Identifiers

CAS 1497-49-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C17H16N2·HCl
Formula Weight 284.78
MDL Number MFCD00012630
Chemical Name or Material N-(5-Anilino-2, 4-pentadienylidene)aniline hydrochloride, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC401250050 SDS Fisher Scientific Acros
401250050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1497-49-0
100.0
C17H16N2·HCl
MFCD00012630
≥97.5%
97.5
98%
284.78
N-(5-Anilino-2, 4-pentadienylidene)aniline hydrochloride, 98%

Specifications

1497-49-0
100.0
Powder
C17H16N2·HCl
284.78
Bordeaux to purple
N-(5-Anilino-2, 4-pentadienylidene)aniline hydrochloride, 98%
97.5
Authentic
98%
Glass bottle
MFCD00012630
5g
≥97.5%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 216-094-3

TSCA : TSCA