Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(5-Aminopentyl)biotinamide trifluoroacetate salt

Chemical Identifiers

Chemical Name or Material N-(5-Aminopentyl)biotinamide trifluoroacetate salt
Formula Weight 442.5
MDL Number MFCD03452814
Synonym Biotin-cadaverine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66382MF

 
alfa aesar™
J66382MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 442.5
Chemical Name or Material N-(5-Aminopentyl)biotinamide trifluoroacetate salt
MDL Number MFCD03452814
Sensitivity Moisture & Light Sensitive
Physical Form Solid
Synonym Biotin-cadaverine
Color White to off-white
Quantity 50mg