Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Pyridyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(4-Pyridyl)thiourea
CAS 164670-44-4
Molecular Formula C6H7N3S
Formula Weight 153.21
MDL Number MFCD00068177
UN Number UN2811
Beilstein 386892
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0992503

 
alfa aesar™
L0992503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0992506

 
alfa aesar™
L0992506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.21
Chemical Name or Material N-(4-Pyridyl)thiourea
Assay 98%
Molecular Formula C6H7N3S
MDL Number MFCD00068177
CAS 164670-44-4
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P301+P310-P330-P501a
Wash thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 181° to 184°C
UN Number UN2811
Beilstein 386892
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 1g

Applications
It is an intermediate of thiazoles.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation.

Safety and Handling

Danger

GHS P Statement
P264-P301+P310-P330-P501a
Wash thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.