Please login to your online account to display your discounted pricing

N-(4-Pyridyl)thiourea, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material N-(4-Pyridyl)thiourea
CAS 164670-44-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7N3S
Formula Weight 153.21
MDL Number MFCD00068177
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02754EA

  SDS
fisher scientific company
SEW02754EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.21
Quantity 10g
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Assay Percent Range 97%
CAS 164670-44-4
Chemical Name or Material N-(4-Pyridyl)thiourea
MDL Number MFCD00068177
Packaging Amber Glass Bottle
Molecular Formula C6H7N3S
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.