missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Maybridge

Chemical Identifiers

CAS 857283-76-2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C15H22N2
Formula Weight 230.35
MDL Number MFCD07772814
Chemical Name or Material N-(4-piperidin-1-ylbenzyl)cyclopropanamine
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC33958CB SDS Fisher Scientific Company
CC33958CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33958DA SDS Fisher Scientific Company
CC33958DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

857283-76-2
100.0
C15H22N2
MFCD07772814
95%
94.5
95%
230.35
N-(4-piperidin-1-ylbenzyl)cyclopropanamine

Specifications

230.35
100.0
857283-76-2
94.5
Amber Glass Bottle
95%
250mg
MFCD07772814
C15H22N2
95%
N-(4-piperidin-1-ylbenzyl)cyclopropanamine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger