Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Phenoxyphenyl)thiourea, 96%

$88.05 - $327.86

Chemical Identifiers

Chemical Name or Material N-(4-Phenoxyphenyl)thiourea
CAS 76839-21-9
Molecular Formula C13H12N2OS
Formula Weight 244.32
MDL Number MFCD00041185
Beilstein 3309384
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1180406

 
alfa aesar™
L1180406
5g Each for $327.86

AAL1180403

 
alfa aesar™
L1180403
1g Each for $88.05
Description & Specifications

Specifications

Formula Weight 244.32
Chemical Name or Material N-(4-Phenoxyphenyl)thiourea
Assay 96%
MDL Number MFCD00041185
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Molecular Formula C13H12N2OS
CAS 76839-21-9
Beilstein 3309384
Health Hazard 3 GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 175° to 177°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.