missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Phenoxyphenyl)thiourea, 96%

Manufacturer:  Alfa Aesar™ L1180403

 View more versions of this product

Catalog No. AAL1180403



Specifications

Specifications

N-(4-Phenoxyphenyl)thiourea
1g
C13H12N2OS
3309384
SSZWMEWZRURHTK-UHFFFAOYSA-N
(4-phenoxyphenyl)thiourea
2760349
96%
175°C to 177°C
76839-21-9
MFCD00041185
1-4-phenoxyphenyl-2-thiourea, 4-phenoxyphenyl thiourea, n-4-phenoxyphenyl thiourea, 1-4-phenoxyphenyl thiourea, amino 4-phenoxyphenyl amino methane-1-thione, 1-4-phenoxyphenyl 2-thiourea, thiourea,n-4-phenoxyphenyl, thiourea, n-4-phenoxyphenyl
C1=CC=C(C=C1)OC2=CC=C(C=C2)NC(=S)N
244.312
244.32

Chemical Identifiers

76839-21-9
244.312
SSZWMEWZRURHTK-UHFFFAOYSA-N
2760349
C1=CC=C(C=C1)OC2=CC=C(C=C2)NC(=S)N
C13H12N2OS
MFCD00041185
1-4-phenoxyphenyl-2-thiourea, 4-phenoxyphenyl thiourea, n-4-phenoxyphenyl thiourea, 1-4-phenoxyphenyl thiourea, amino 4-phenoxyphenyl amino methane-1-thione, 1-4-phenoxyphenyl 2-thiourea, thiourea,n-4-phenoxyphenyl, thiourea, n-4-phenoxyphenyl
(4-phenoxyphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f-P308+P313-P330-P501a
Wear protective clothing.
IF exposed or concerned: Get medical advice/attention.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents