missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(4-Nitrobenzoyl)-beta-alanine, TCI America™

$68.75 - $210.84

Chemical Identifiers

CAS 59642-21-6
Molecular Formula C10H10N2O5
Molecular Weight (g/mol) 238.199
MDL Number MFCD00009794
InChI Key PDTLZWITKYGYDN-UHFFFAOYSA-N
Synonym 3-(4-Nitrobenzamido)propionic Acid
PubChem CID 591374
SMILES C1=CC(=CC=C1C(=O)NCCC(=O)O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09425G View Documents TCI America
N09425G
5g Each for $68.75
 
N094225G View Documents TCI America
N094225G
25g Each for $210.84
 
Specifications

Chemical Identifiers

59642-21-6
238.199
PDTLZWITKYGYDN-UHFFFAOYSA-N
591374
C10H10N2O5
MFCD00009794
3-(4-Nitrobenzamido)propionic Acid
C1=CC(=CC=C1C(=O)NCCC(=O)O)[N+](=O)[O-]

Specifications

Yellow
5g
C10H10N2O5
3-(4-Nitrobenzamido)propionic Acid
C1=CC(=CC=C1C(=O)NCCC(=O)O)[N+](=O)[O-]
591374
Crystal-Powder at 20°C
N-(4-Nitrobenzoyl)-beta-alanine
165°C
59642-21-6
MFCD00009794
PDTLZWITKYGYDN-UHFFFAOYSA-N
238.199
238.20
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No