Please login to your online account to display your discounted pricing

N-[[(4-Methylphenyl)sulfonyl]oxy]ethane -1,2-diamine, ≥95%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01735EA

 View more versions of this product

Catalog No. SB01735EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 230.29
Quantity 10g
Assay 95%
CAS Min % 95.0
Molecular Formula C9H14N2O3S
Linear Formula CH3C6H4SO2ONH(CH2)2NH2
CAS 175205-36-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Amber glass bottle
Chemical Name or Material N-[[(4-Methylphenyl)sulfonyl]oxy]ethane -1, 2-diamine
Assay Percent Range 95%+
MDL Number MFCD00276588
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-[[(4-Methylphenyl)sulfonyl]oxy]ethane -1, 2-diamine
CAS 175205-36-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C9H14N2O3S
Linear Formula CH3C6H4SO2ONH(CH2)2NH2
Formula Weight 230.29
MDL Number MFCD00276588
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents