missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(4-Methyl-2-pyridyl)acetamide, TCI America™

$35.14 - $103.89

Chemical Identifiers

CAS 5327-32-2
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00234432
InChI Key QGZHGSGLCZEGHA-UHFFFAOYSA-N
Synonym 2-Acetamido-4-methylpyridine
PubChem CID 220485
IUPAC Name N-(4-methylpyridin-2-yl)acetamide
SMILES CC1=CC(=NC=C1)NC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27875G View Documents TCI America
M27875G
5g Each for $103.89
 
M27871G View Documents TCI America
M27871G
1g Each for $35.14
 
Specifications

Chemical Identifiers

5327-32-2
150.181
QGZHGSGLCZEGHA-UHFFFAOYSA-N
220485
CC1=CC(=NC=C1)NC(=O)C
C8H10N2O
MFCD00234432
2-Acetamido-4-methylpyridine
N-(4-methylpyridin-2-yl)acetamide

Specifications

White
5g
C8H10N2O
2-Acetamido-4-methylpyridine
CC1=CC(=NC=C1)NC(=O)C
150.181
150.18
≥98.0% (GC)
104°C
5327-32-2
MFCD00234432
QGZHGSGLCZEGHA-UHFFFAOYSA-N
N-(4-methylpyridin-2-yl)acetamide
220485
Crystal-Powder at 20°C
N-(4-Methyl-2-pyridyl)acetamide
Safety and Handling

Safety and Handling

TSCA : No