missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Methoxyphenyl)thiourea, 96%

$334.64 - $334.64

Chemical Identifiers

CAS 2293-07-4
Molecular Formula C8H10N2OS
Molecular Weight (g/mol) 182.24
MDL Number MFCD00004936
InChI Key SRYLJBWDZZMDSK-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl thiourea, 4-methoxyphenyl thiourea, n-4-methoxyphenyl thiourea, 1-4-methoxyphenyl-2-thiourea, p-anisylthiourea, n-p-anisylthiourea, p-methoxyphenyl thiourea, thiourea, 4-methoxyphenyl, n-p-methoxyphenyl thiourea, 4-methoxyphenyl thiourea
PubChem CID 667549
IUPAC Name (4-methoxyphenyl)thiourea
SMILES COC1=CC=C(C=C1)NC(=S)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0073914 View Documents Alfa Aesar™
L0073914
25g Each for $334.64
 
Specifications

Chemical Identifiers

2293-07-4
182.24
SRYLJBWDZZMDSK-UHFFFAOYSA-N
667549
COC1=CC=C(C=C1)NC(=S)N
C8H10N2OS
MFCD00004936
1-4-methoxyphenyl thiourea, 4-methoxyphenyl thiourea, n-4-methoxyphenyl thiourea, 1-4-methoxyphenyl-2-thiourea, p-anisylthiourea, n-p-anisylthiourea, p-methoxyphenyl thiourea, thiourea, 4-methoxyphenyl, n-p-methoxyphenyl thiourea, 4-methoxyphenyl thiourea
(4-methoxyphenyl)thiourea

Specifications

Odorless
25g
C8H10N2OS
UN2811
1-4-methoxyphenyl thiourea, 4-methoxyphenyl thiourea, n-4-methoxyphenyl thiourea, 1-4-methoxyphenyl-2-thiourea, p-anisylthiourea, n-p-anisylthiourea, p-methoxyphenyl thiourea, thiourea, 4-methoxyphenyl, n-p-methoxyphenyl thiourea, 4-methoxyphenyl thiourea
COC1=CC=C(C=C1)NC(=S)N
182.24
182.25
N-(4-Methoxyphenyl)thiourea
~212°C
2293-07-4
MFCD00004936
777238
SRYLJBWDZZMDSK-UHFFFAOYSA-N
(4-methoxyphenyl)thiourea
667549
96%
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YT6825000