Please login to your online account to display your discounted pricing

N-(4-Methoxyphenyl)-p-phenylenediamine Hydrochloride 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  413890250

 View more versions of this product

Catalog No. AC413890250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 250.73
Quantity 25g
Color Gray-blue
Beilstein 13, III, 1161
Merck Index 15, 6072
EINECS Number 222-652-7
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C13H14N2O·HCl
MDL Number MFCD00012989
CAS 3566-44-7

Specifications

Chemical Name or Material N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride
Name Note 95%
CAS 3566-44-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H14N2O·HCl
Formula Weight 250.73
MDL Number MFCD00012989