Please login to your online account to display your discounted pricing

N-(4-Methoxyphenyl)-p-phenylenediamine Hydrochloride 95%, ACROS Organics™

$197.39 - $197.39

Chemical Identifiers

Chemical Name or Material N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride
Name Note 95%
CAS 3566-44-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H14N2O·HCl
Formula Weight 250.73
MDL Number MFCD00012989
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC413890250

  SDS
thermo fisher scientific
413890250
Glass bottle 25g Each for $197.39
Description & Specifications

Specifications

Assay ≥94%
Formula Weight 250.73
Physical Form Crystalline Powder
Quantity 25g
Color Gray-blue
Beilstein 13, III, 1161
Merck Index 15, 6072
TSCA TSCA
EINECS Number 222-652-7
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride
CAS 3566-44-7
Assay Percent Range 95%
Molecular Formula C13H14N2O·HCl
Name Note 95%
MDL Number MFCD00012989