Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide, 97%

Chemical Identifiers

Chemical Name or Material N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide
CAS 180631-81-6
Formula Weight 308.3
MDL Number MFCD00414861
Beilstein 2764039
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58131MC

 
alfa aesar™
H58131MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH58131ME

 
alfa aesar™
H58131ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 308.3
Chemical Name or Material N-(4-Methoxyphenyl)-3-nitrobenzenesulfonamide
Physical Form Solid
Assay 97%
MDL Number MFCD00414861
CAS 180631-81-6
Beilstein 2764039
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Pink
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.