missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Methoxybenzylidene)aniline, 98%

$43.49 - $135.24

Chemical Identifiers

CAS 836-41-9
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.264
MDL Number MFCD00025836
InChI Key MSWPGMRTURVKRJ-UHFFFAOYSA-N
Synonym n-4-methoxybenzylidene aniline, e-n-4-methoxybenzylidene aniline, n-p-anisal aniline, n-p-anisylideneaniline, 1-4-methoxyphenyl-n-phenylmethanimine, 4-methoxybenzyliden-anilin, 4-methoxybenzylideneaniline, 1e-1-4-methoxyphenyl-n-phenylmethanimine, p-methoxybenzylidene-phenyl-amine, n-4-methoxyphenyl methylidene aniline #
PubChem CID 98633
IUPAC Name 1-(4-methoxyphenyl)-N-phenylmethanimine
SMILES COC1=CC=C(C=C1)C=NC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0195106 View Documents Alfa Aesar™
L0195106
5g Each for $43.49
 
AAL0195114 View Documents Alfa Aesar™
L0195114
25g Each for $135.24
 
Specifications

Chemical Identifiers

836-41-9
211.264
MSWPGMRTURVKRJ-UHFFFAOYSA-N
98633
COC1=CC=C(C=C1)C=NC2=CC=CC=C2
C14H13NO
MFCD00025836
n-4-methoxybenzylidene aniline, e-n-4-methoxybenzylidene aniline, n-p-anisal aniline, n-p-anisylideneaniline, 1-4-methoxyphenyl-n-phenylmethanimine, 4-methoxybenzyliden-anilin, 4-methoxybenzylideneaniline, 1e-1-4-methoxyphenyl-n-phenylmethanimine, p-methoxybenzylidene-phenyl-amine, n-4-methoxyphenyl methylidene aniline #
1-(4-methoxyphenyl)-N-phenylmethanimine

Specifications

61°C to 64°C
836-41-9
MFCD00025836
n-4-methoxybenzylidene aniline, e-n-4-methoxybenzylidene aniline, n-p-anisal aniline, n-p-anisylideneaniline, 1-4-methoxyphenyl-n-phenylmethanimine, 4-methoxybenzyliden-anilin, 4-methoxybenzylideneaniline, 1e-1-4-methoxyphenyl-n-phenylmethanimine, p-methoxybenzylidene-phenyl-amine, n-4-methoxyphenyl methylidene aniline #
COC1=CC=C(C=C1)C=NC2=CC=CC=C2
211.264
211.26
N-(4-Methoxybenzylidene)aniline
5g
C14H13NO
642337
MSWPGMRTURVKRJ-UHFFFAOYSA-N
1-(4-methoxyphenyl)-N-phenylmethanimine
98633
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger