Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Iodophenyl)maleamic acid, 97%

$28.62 - $406.36

Chemical Identifiers

Chemical Name or Material N-(4-Iodophenyl)maleamic acid
CAS 17280-49-8
Molecular Formula C10H8INO3
Formula Weight 317.08
MDL Number MFCD00020473
Synonym 4-(4-Iodoanilino)-4-oxobut-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2521003

 
alfa aesar™
B2521003
1g Each for $28.62

AAB2521006

 
alfa aesar™
B2521006
5g Each for $98.05

AAB2521014

 
alfa aesar™
B2521014
25g Each for $406.36
Description & Specifications

Specifications

Formula Weight 317.08
Chemical Name or Material N-(4-Iodophenyl)maleamic acid
Assay 97%
Molecular Formula C10H8INO3
MDL Number MFCD00020473
CAS 17280-49-8
Synonym 4-(4-Iodoanilino)-4-oxobut-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 193° to 194°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.