missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(4-Hydroxyphenyl)thiourea, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1253703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-(4-Hydroxyphenyl)thiourea | |
∽210°C (decomposition) | |
C7H8N2OS | |
1 g | |
1-4-hydroxyphenyl-2-thiourea, 1-4-hydroxyphenyl thiourea, 4-hydroxyphenyl thiourea, p-hydroxyphenylthiourea, usaf b-75, n-4-hydroxyphenyl thiourea, urea, 1-p-hydroxyphenyl-2-thio, thiourea, 4-hydroxyphenyl, 4-hydroxy-phenyl-thiourea, 4-aminothioxomethyl amino phenol | |
C1=CC(=CC=C1NC(=S)N)O | |
168.214 | |
168.22 |
1520-27-0 | |
Odorless | |
MFCD00041186 | |
1102429 | |
QICKOOCQSYZYQB-UHFFFAOYSA-N | |
(4-hydroxyphenyl)thiourea | |
2759331 | |
≥98% |
Chemical Identifiers
1520-27-0 | |
168.214 | |
QICKOOCQSYZYQB-UHFFFAOYSA-N | |
2759331 | |
C1=CC(=CC=C1NC(=S)N)O |
C7H8N2OS | |
MFCD00041186 | |
1-4-hydroxyphenyl-2-thiourea, 1-4-hydroxyphenyl thiourea, 4-hydroxyphenyl thiourea, p-hydroxyphenylthiourea, usaf b-75, n-4-hydroxyphenyl thiourea, urea, 1-p-hydroxyphenyl-2-thio, thiourea, 4-hydroxyphenyl, 4-hydroxy-phenyl-thiourea, 4-aminothioxomethyl amino phenol | |
(4-hydroxyphenyl)thiourea |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : YT5250000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only