Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Hydroxyphenyl)phthalimide, 98+%

$49.11 - $164.24

Chemical Identifiers

Chemical Name or Material N-(4-Hydroxyphenyl)phthalimide
CAS 7154-85-0
Molecular Formula C14H9NO3
Formula Weight 239.23
MDL Number MFCD00023050
Beilstein 384177
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1307904

  SDS
alfa aesar™
L1307904
2g Each for $49.11

AAL1307909

  SDS
alfa aesar™
L1307909
10g Each for $164.24
Description & Specifications

Specifications

Formula Weight 239.23
Chemical Name or Material N-(4-Hydroxyphenyl)phthalimide
Molecular Formula C14H9NO3
CAS 7154-85-0
Beilstein 384177
Health Hazard 1 Warning
Assay ≥98%
MDL Number MFCD00023050
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 295° to 297°C
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.