missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Hydroxyphenyl)phthalimide, 98+%

Manufacturer:  Alfa Aesar™ L1307904

 View more versions of this product

Catalog No. AAL1307904



Specifications

Specifications

N-(4-Hydroxyphenyl)phthalimide
2g
C14H9NO3
384177
PHHOJUGXERSDJH-UHFFFAOYSA-N
2-(4-hydroxyphenyl)isoindole-1,3-dione
252281
≥98%
295°C to 297°C
7154-85-0
MFCD00023050
n-4-hydroxyphenyl phthalimide, 2-4-hydroxyphenyl-1h-isoindole-1,3 2h-dione, 2-4-hydroxyphenyl isoindoline-1,3-dione, 2-4-hydroxyphenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-4-hydroxyphenyl isoindole-1,3-dione, cbmicro_029453, n-p-hydroxyphenyl phthalimide, n-4-hydroxyphenyl phthalamide, 2-4-hydroxy-phenyl-isoindole-1,3-dione, 2-4-hydroxyphenyl benzo c azoline-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)C3=CC=C(C=C3)O
239.23
239.23

Chemical Identifiers

7154-85-0
239.23
PHHOJUGXERSDJH-UHFFFAOYSA-N
252281
C1=CC=C2C(=C1)C(=O)N(C2=O)C3=CC=C(C=C3)O
C14H9NO3
MFCD00023050
n-4-hydroxyphenyl phthalimide, 2-4-hydroxyphenyl-1h-isoindole-1,3 2h-dione, 2-4-hydroxyphenyl isoindoline-1,3-dione, 2-4-hydroxyphenyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-4-hydroxyphenyl isoindole-1,3-dione, cbmicro_029453, n-p-hydroxyphenyl phthalimide, n-4-hydroxyphenyl phthalamide, 2-4-hydroxy-phenyl-isoindole-1,3-dione, 2-4-hydroxyphenyl benzo c azoline-1,3-dione
2-(4-hydroxyphenyl)isoindole-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents