Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Hydroxyphenyl)glycine, 98%

$92.66 - $280.15

Chemical Identifiers

Chemical Name or Material N-(4-Hydroxyphenyl)glycine
CAS 122-87-2
Molecular Formula C8H9NO3
Formula Weight 167.16
MDL Number MFCD00002689
Merck Index 14,4837
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1684309

 
alfa aesar™
A1684309
10g Each for $92.66

AAA1684318

 
alfa aesar™
A1684318
50g Each for $280.15
Description & Specifications

Specifications

Formula Weight 167.16
Chemical Name or Material N-(4-Hydroxyphenyl)glycine
TSCA TSCA
Assay 98%
Molecular Formula C8H9NO3
MDL Number MFCD00002689
CAS 122-87-2
Merck Index 14,4837
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈240°C (dec.)
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.