Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorophenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N-(4-Fluorophenyl)thiourea
CAS 459-05-2
Molecular Formula C7H7FN2S
Formula Weight 170.21
MDL Number MFCD00041180
UN Number UN2811
Beilstein 2803645
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1115903

  SDS
alfa aesar™
L1115903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1115906

  SDS
alfa aesar™
L1115906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.21
Chemical Name or Material N-(4-Fluorophenyl)thiourea
Assay 99%
Molecular Formula C7H7FN2S
MDL Number MFCD00041180
CAS 459-05-2
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Melting Point 162° to 166°C
UN Number UN2811
Beilstein 2803645
Quantity 1g
Odor Odorless
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.