missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorophenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ L1115903

 View more versions of this product

Catalog No. AAL1115903



Specifications

Specifications

N-(4-Fluorophenyl)thiourea
162°C to 166°C
459-05-2
MFCD00041180
2803645
BRWKXKNZRVALNZ-UHFFFAOYSA-N
(4-fluorophenyl)thiourea
693061
99%
Odorless
1g
C7H7FN2S
UN2811
1-4-fluorophenyl thiourea, 1-4-fluorophenyl-2-thiourea, 4-fluorophenyl thiourea, n-4-fluorophenyl thiourea, p-fluorophenylthiourea, 4-fluoro-phenyl-thiourea, thiourea, 4-fluorophenyl, amino 4-fluorophenyl amino methane-1-thione, pubchem10059, acmc-1ajjb
C1=CC(=CC=C1NC(=S)N)F
170.205
170.21

Chemical Identifiers

459-05-2
170.205
BRWKXKNZRVALNZ-UHFFFAOYSA-N
693061
C1=CC(=CC=C1NC(=S)N)F
C7H7FN2S
MFCD00041180
1-4-fluorophenyl thiourea, 1-4-fluorophenyl-2-thiourea, 4-fluorophenyl thiourea, n-4-fluorophenyl thiourea, p-fluorophenylthiourea, 4-fluoro-phenyl-thiourea, thiourea, 4-fluorophenyl, amino 4-fluorophenyl amino methane-1-thione, pubchem10059, acmc-1ajjb
(4-fluorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents