Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorophenyl)maleamic acid, 96%

$35.09 - $105.30

Chemical Identifiers

Chemical Name or Material N-(4-Fluorophenyl)maleamic acid
CAS 60252-79-1
Molecular Formula C10H8FNO3
Formula Weight 209.18
MDL Number MFCD00082643
Beilstein 2842921
Synonym Maleic acid mono(4-fluorophenyl)amide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1039403

  SDS
alfa aesar™
L1039403
1g Each for $35.09

AAL1039406

  SDS
alfa aesar™
L1039406
5g Each for $105.30
Description & Specifications

Specifications

Formula Weight 209.18
Chemical Name or Material N-(4-Fluorophenyl)maleamic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 96%
Molecular Formula C10H8FNO3
MDL Number MFCD00082643
Beilstein 2842921
Quantity 1g
Synonym Maleic acid mono(4-fluorophenyl)amide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 207° to 209°C
CAS 60252-79-1
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.