missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(4-Fluorophenyl)maleamic acid, 96%

Manufacturer:  Alfa Aesar™ L1039406

 View more versions of this product

Catalog No. AAL1039406



Specifications

Specifications

N-(4-Fluorophenyl)maleamic acid
5g
C10H8FNO3
2842921
NRDZVHHPNZDWRA-WAYWQWQTSA-N
(Z)-4-(4-fluoroanilino)-4-oxobut-2-enoic acid
1551046
96%
207°C to 209°C
60252-79-1
MFCD00082643
z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, 2z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamic acid, 2z-3-4-fluorophenyl carbamoyl prop-2-enoic acid, 2e-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamidic acid, z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2-butenoic acid,4-4-fluorophenyl amino-4-oxo-, 2z
C1=CC(=CC=C1NC(=O)C=CC(=O)O)F
209.176
209.18

Chemical Identifiers

60252-79-1
209.176
NRDZVHHPNZDWRA-WAYWQWQTSA-N
1551046
C1=CC(=CC=C1NC(=O)C=CC(=O)O)F
C10H8FNO3
MFCD00082643
z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, 2z-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamic acid, 2z-3-4-fluorophenyl carbamoyl prop-2-enoic acid, 2e-4-4-fluorophenyl amino-4-oxobut-2-enoic acid, n-4-fluorophenyl maleamidic acid, z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2z-4-4-fluoroanilino-4-oxobut-2-enoic acid, 2-butenoic acid,4-4-fluorophenyl amino-4-oxo-, 2z
(Z)-4-(4-fluoroanilino)-4-oxobut-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents